Product Details

SNP ID
rs137914321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128259599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGATTTCCAGTGTGTATGTGCC[A/G]GGCATGAGCTTCTTCTTGGCCGTGT
Phenotype
MIM: 612570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBN2 PubMed Links

Gene Details

Gene
FBN2
Gene Name
fibrillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001999.3 9197 Silent Mutation CCC,CCT P2865P NP_001990.2
XM_017009228.1 9197 Silent Mutation CCC,CCT P2814P XP_016864717.1

View Full Product Details