Product Details

SNP ID
rs138344347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:57482019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGGTGAGGAGGATGCGGCTGCC[C/T]TGACCATCTCCTGCTCAAAGTCCAG
Phenotype
MIM: 614835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTBL2 PubMed Links

Gene Details

Gene
ACTBL2
Gene Name
actin, beta like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017992.3 791 Missense Mutation AAG,AGG K230R NP_001017992.1

View Full Product Details