Product Details

SNP ID
rs138739318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:142001549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACTGCTTCCTTGCACTTATTTT[A/G]GATATTGAGAGTTTGTGCCTTCACT
Phenotype
MIM: 601798 MIM: 605675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNPDA1 PubMed Links

Gene Details

Gene
GNPDA1
Gene Name
glucosamine-6-phosphate deaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005471.4 1417 UTR 3 NP_005462.1
XM_005268348.1 1417 UTR 3 XP_005268405.1
XM_006714747.2 1417 UTR 3 XP_006714810.1
XM_011537537.1 1417 UTR 3 XP_011535839.1
Gene
RNF14
Gene Name
ring finger protein 14
There are no transcripts associated with this gene.

View Full Product Details