Product Details

SNP ID
rs138792608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64769166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGATGAGCAACATCTACATCC[A/C]GGAGCCTCCCACGAATGGGAAGGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CWC27 PubMed Links

Gene Details

Gene
CWC27
Gene Name
CWC27 spliceosome associated protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297644.1 249 Missense Mutation CAG,CCG Q7P NP_001284573.1
NM_001297645.1 249 Missense Mutation CAG,CCG Q7P NP_001284574.1
NM_001318000.1 249 Missense Mutation CAG,CCG Q7P NP_001304929.1
NM_005869.3 249 Missense Mutation CAG,CCG Q7P NP_005860.2
XM_011543095.2 249 Missense Mutation CAG,CCG Q7P XP_011541397.2
XM_011543096.1 249 Missense Mutation CAG,CCG Q7P XP_011541398.1
XM_017008943.1 249 Missense Mutation CAG,CCG Q7P XP_016864432.1
Gene
SREK1IP1
Gene Name
SREK1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173829.3 249 Intron NP_776190.1
XM_011543344.2 249 Intron XP_011541646.1

View Full Product Details