Product Details

SNP ID
rs138905856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150053569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGCAGAGACTAAGAGGTCCTGT[A/G]AGATTCTTAGAGGAGCCATCCTGCT
Phenotype
MIM: 164770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSF1R PubMed Links

Gene Details

Gene
CSF1R
Gene Name
colony stimulating factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288705.1 3711 UTR 3 NP_001275634.1
NM_005211.3 3711 UTR 3 NP_005202.2
Gene
HMGXB3
Gene Name
HMG-box containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014983.2 3711 Intron NP_055798.2
XM_006714765.2 3711 Intron XP_006714828.1

View Full Product Details