Product Details

SNP ID
rs139459678
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:144160553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGCAGTGAGAATGATTCCTAC[C/T]ATTCCTCTGTAAACAACAAGGAAAA
Phenotype
MIM: 611483
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
YIPF5 PubMed Links

Gene Details

Gene
YIPF5
Gene Name
Yip1 domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024947.3 808 Missense Mutation ATA,ATG I206M NP_001020118.1
NM_001271732.1 808 Missense Mutation ATA,ATG I152M NP_001258661.1
NM_030799.8 808 Missense Mutation ATA,ATG I206M NP_110426.4

View Full Product Details