Product Details

SNP ID
rs139484791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:109337295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGGATCAGGCTCAGAGGAAGGA[A/G]CTGCAGATGCTTCAATAACCGCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC285638 PubMed Links

Gene Details

Gene
LOC285638
Gene Name
uncharacterized LOC285638
There are no transcripts associated with this gene.

Gene
PJA2
Gene Name
praja ring finger ubiquitin ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014819.4 2359 Missense Mutation GCT,GTT A688V NP_055634.3
XM_017010099.1 2359 Missense Mutation GCT,GTT A688V XP_016865588.1

View Full Product Details