Product Details

SNP ID
rs140064341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157181092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTGTTAACCAAAGCCAGCCTG[G/T]CATACTTTGAAGATCGTCATGGGGT
Phenotype
MIM: 186973
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ITK PubMed Links

Gene Details

Gene
ITK
Gene Name
IL2 inducible T-cell kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005546.3 197 Missense Mutation GCA,TCA A39S NP_005537.3
XM_017009443.1 197 Intron XP_016864932.1

View Full Product Details