Product Details

SNP ID
rs140118369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150054185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCTCACTGCTGCTGCTGCCGC[C/T]GCCACCGCTTCTGCTGCTGCTCGGC
Phenotype
MIM: 164770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSF1R PubMed Links

Gene Details

Gene
CSF1R
Gene Name
colony stimulating factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288705.1 3095 Missense Mutation AGC,GGC S935G NP_001275634.1
NM_005211.3 3095 Missense Mutation AGC,GGC S935G NP_005202.2
Gene
HMGXB3
Gene Name
HMG-box containing 3
There are no transcripts associated with this gene.

View Full Product Details