Product Details
- SNP ID
-
rs140135215
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:154596067 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTCCTCCTCCACCCGCCCTCACC[A/G]GGTGAGTCCCTCTACATCCCTGTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3141
PubMed Links
Gene Details
- Gene
- MIR3141
- Gene Name
- microRNA 3141
There are no transcripts associated with this gene.
View Full Product Details