Product Details

SNP ID
rs140373868
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148987022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGGAAAATGAAATATTAATTC[C/T]CCAGCTAATGTATTTCATGGTAGTT
Phenotype
MIM: 608206
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SH3TC2 PubMed Links

Gene Details

Gene
SH3TC2
Gene Name
SH3 domain and tetratricopeptide repeats 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024577.3 21708 UTR 3 NP_078853.2

View Full Product Details