Product Details

SNP ID
rs140398806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141051595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTGGCAGAGGAAATGGAGAGCG[A/G]CTCGTTTGTGGCCAACGTAGCTAAG
Phenotype
MIM: 606327
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101926905 PubMed Links

Gene Details

Gene
LOC101926905
Gene Name
uncharacterized LOC101926905
There are no transcripts associated with this gene.

Gene
PCDHB1
Gene Name
protocadherin beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013340.3 220 Missense Mutation GAC,GGC D42G NP_037472.2

View Full Product Details