Product Details

SNP ID
rs140680370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126780508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTTTCAGCGGGGTTAAAAAAT[A/G]AGAATAATGTGTATTAGAATCAATG
Phenotype
MIM: 150340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMNB1 PubMed Links

Gene Details

Gene
LMNB1
Gene Name
lamin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198557.1 Intron NP_001185486.1
NM_005573.3 Intron NP_005564.1
Gene
LOC102723557
Gene Name
uncharacterized LOC102723557
There are no transcripts associated with this gene.

View Full Product Details