Product Details

SNP ID
rs140930836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37292934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACATATCTTATCAATAATACAT[C/G]TATACAATCCAAAAGGTGCAGTGGC
Phenotype
MIM: 606694
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUP155 PubMed Links

Gene Details

Gene
NUP155
Gene Name
nucleoporin 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278312.1 4131 Missense Mutation CAT,GAT H1264D NP_001265241.1
NM_004298.3 4131 Missense Mutation CAT,GAT H1269D NP_004289.1
NM_153485.2 4131 Missense Mutation CAT,GAT H1328D NP_705618.1
XM_011514165.2 4131 Intron XP_011512467.1

View Full Product Details