Product Details

SNP ID
rs141507307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132815014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGTCTTGCTGCCAGGCGCCCC[A/G]TACAGGTGGCCGCCACCATCCTCAG
Phenotype
MIM: 611179
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHROOM1 PubMed Links

Gene Details

Gene
SHROOM1
Gene Name
shroom family member 1
There are no transcripts associated with this gene.

Gene
SOWAHA
Gene Name
sosondowah ankyrin repeat domain family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175873.5 1691 Missense Mutation ATA,GTA I465V NP_787069.4

View Full Product Details