Product Details

SNP ID
rs141582754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177430966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCGAAGAGCTGCGGCTCAGCCT[C/T]GGTGAGGCCTGGGCAGAGACTGGGG
Phenotype
MIM: 600869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRK6 PubMed Links

Gene Details

Gene
GRK6
Gene Name
G protein-coupled receptor kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004105.2 307 Silent Mutation CTC,CTT L49L NP_001004105.1
NM_001004106.2 307 Silent Mutation CTC,CTT L49L NP_001004106.1
NM_002082.3 307 Silent Mutation CTC,CTT L49L NP_002073.2
XM_006714858.3 307 Intron XP_006714921.1
XM_006714859.2 307 Silent Mutation CTC,CTT L17L XP_006714922.1
XM_011534537.2 307 Missense Mutation TCG,TTG S48L XP_011532839.1
XM_011534538.1 307 Intron XP_011532840.1
Gene
PRR7-AS1
Gene Name
PRR7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details