Product Details

SNP ID
rs142045369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:41909782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGACATATTTTGAGGAGAACAAG[A/C]TAGTAGATGAAGATTTTCCTGAAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C5orf51 PubMed Links

Gene Details

Gene
C5orf51
Gene Name
chromosome 5 open reading frame 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175921.5 397 Missense Mutation ATA,CTA I82L NP_787117.3
XM_005248289.4 397 Missense Mutation ATA,CTA I82L XP_005248346.1
XM_011514032.2 397 Missense Mutation ATA,CTA I9L XP_011512334.1

View Full Product Details