Product Details
- SNP ID
-
rs142122458
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141122674 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCACCACCTAGGTCTGGCACGGTC[A/G]TGGTTCGAATCCTGATCATGGACAT
- Phenotype
-
MIM: 606330
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHB4
PubMed Links
Gene Details
- Gene
- PCDHB4
- Gene Name
- protocadherin beta 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018938.3 |
925 |
Missense Mutation |
ATG,GTG |
M226V |
NP_061761.1 |
View Full Product Details