Product Details

SNP ID
rs142691743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54455814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCAAAAATCATTTTGAGGCAC[C/T]TCATAGAGATTCCAGTGCGTTACCA
Phenotype
MIM: 604624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPB3 PubMed Links

Gene Details

Gene
HSPB3
Gene Name
heat shock protein family B (small) member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006308.2 214 Missense Mutation CTC,TTC L9F NP_006299.1

View Full Product Details