Product Details
- SNP ID
-
rs142916882
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138286570 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCTCTTCTGGGTGTCCAAAGGGA[C/T]GATGGGCTTCTTCAGAAAGAAGTTA
- Phenotype
-
MIM: 157680
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC25C
PubMed Links
Gene Details
- Gene
- CDC25C
- Gene Name
- cell division cycle 25C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287582.1 |
1521 |
Missense Mutation |
ATC,GTC |
I363V |
NP_001274511.1 |
NM_001287583.1 |
1521 |
Missense Mutation |
ATC,GTC |
I441V |
NP_001274512.1 |
NM_001318098.1 |
1521 |
Missense Mutation |
ATC,GTC |
I380V |
NP_001305027.1 |
NM_001790.4 |
1521 |
Missense Mutation |
ATC,GTC |
I363V |
NP_001781.2 |
NM_022809.3 |
1521 |
Missense Mutation |
ATC,GTC |
I290V |
NP_073720.1 |
XM_005272145.3 |
1521 |
Missense Mutation |
ATC,GTC |
I407V |
XP_005272202.1 |
XM_006714739.3 |
1521 |
Missense Mutation |
ATC,GTC |
I365V |
XP_006714802.1 |
XM_011543759.2 |
1521 |
Missense Mutation |
ATC,GTC |
I387V |
XP_011542061.1 |
XM_011543760.2 |
1521 |
Missense Mutation |
ATC,GTC |
I386V |
XP_011542062.1 |
XM_011543761.2 |
1521 |
Missense Mutation |
ATC,GTC |
I368V |
XP_011542063.1 |
XM_011543762.1 |
1521 |
Missense Mutation |
ATC,GTC |
I290V |
XP_011542064.1 |
XM_011543763.1 |
1521 |
Missense Mutation |
ATC,GTC |
I252V |
XP_011542065.1 |
XM_011543764.1 |
1521 |
Missense Mutation |
ATC,GTC |
I201V |
XP_011542066.1 |
XM_017010101.1 |
1521 |
Missense Mutation |
ATC,GTC |
I329V |
XP_016865590.1 |
View Full Product Details