Product Details

SNP ID
rs142950208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:152391944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCTTTCCAGTCACGTTCTTGG[C/T]ATGAACTAGTGAAGGGGCATGAGGC
Phenotype
MIM: 605108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NMUR2 PubMed Links

Gene Details

Gene
NMUR2
Gene Name
neuromedin U receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020167.4 1661 UTR 3 NP_064552.3
XM_011537670.2 1661 Intron XP_011535972.1

View Full Product Details