Product Details

SNP ID
rs143315247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:181155460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAATTTCCCCTACTGTGGCTTGA[A/G]GAAGGTGAACCATTTCTTCTGTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2V2 PubMed Links

Gene Details

Gene
OR2V2
Gene Name
olfactory receptor family 2 subfamily V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206880.1 Intron NP_996763.1

View Full Product Details