Product Details

SNP ID
rs143433399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180845604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGACAATTGAAGTACTGTATTCT[A/G]CTTCCTTCTCCATGTCTCTAGGCCT
Phenotype
MIM: 610281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00847 PubMed Links

Gene Details

Gene
LINC00847
Gene Name
long intergenic non-protein coding RNA 847
There are no transcripts associated with this gene.

Gene
ZFP62
Gene Name
ZFP62 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172638.1 3031 Intron NP_001166109.1
NM_152283.4 3031 Intron NP_689496.4
XM_017009710.1 3031 Intron XP_016865199.1
XM_017009711.1 3031 Intron XP_016865200.1
XM_017009712.1 3031 Intron XP_016865201.1
XM_017009713.1 3031 Intron XP_016865202.1
XM_017009714.1 3031 Intron XP_016865203.1
XM_017009715.1 3031 Intron XP_016865204.1
XM_017009716.1 3031 UTR 3 XP_016865205.1
XM_017009717.1 3031 Intron XP_016865206.1
XM_017009718.1 3031 Intron XP_016865207.1
XM_017009719.1 3031 Intron XP_016865208.1
XM_017009720.1 3031 Intron XP_016865209.1
XM_017009721.1 3031 Intron XP_016865210.1

View Full Product Details