Product Details

SNP ID
rs143577894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:33944837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGTCCATGCCCTTCCCTCTCAC[A/G]CTGTTGTCTGGGTCCCCTCCTGGGG
Phenotype
MIM: 609445 MIM: 606202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RXFP3 PubMed Links

Gene Details

Gene
RXFP3
Gene Name
relaxin/insulin like family peptide receptor 3
There are no transcripts associated with this gene.

Gene
SLC45A2
Gene Name
solute carrier family 45 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012509.3 1496 Intron NP_001012527.1
NM_001297417.2 1496 Intron NP_001284346.2
NM_016180.4 1496 Silent Mutation NP_057264.3

View Full Product Details