Product Details

SNP ID
rs144074026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:16474506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTTATCTCTGACAACACAGTG[G/T]TGTGTATAAAGTTCATTTCCATGCT
Phenotype
MIM: 613114 MIM: 608694
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM134B PubMed Links

Gene Details

Gene
FAM134B
Gene Name
family with sequence similarity 134 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034850.2 1875 UTR 3 NP_001030022.1
NM_019000.4 1875 UTR 3 NP_061873.2
XM_011514053.2 1875 UTR 3 XP_011512355.1
XM_011514054.2 1875 UTR 3 XP_011512356.1
XM_011514055.2 1875 UTR 3 XP_011512357.1
Gene
ZNF622
Gene Name
zinc finger protein 622
There are no transcripts associated with this gene.

View Full Product Details