Product Details

SNP ID
rs145206191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40998672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGCTGGTCAAATTGAGAACAA[C/T]GGCATCTAAAGTTAATAGGAGACCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MROH2B PubMed Links

Gene Details

Gene
MROH2B
Gene Name
maestro heat like repeat family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173489.4 5067 Missense Mutation ATT,GTT I1531V NP_775760.3
XM_011513952.1 5067 Missense Mutation ATT,GTT I1531V XP_011512254.1
XM_011513953.1 5067 Missense Mutation ATT,GTT I1469V XP_011512255.1

View Full Product Details