Product Details

SNP ID
rs145283175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179643436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGGAAGAGCCTCACTTTTTC[A/G]ACAGGAAAGTGTAGATCTCAAGGTT
Phenotype
MIM: 601035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf60 PubMed Links

Gene Details

Gene
C5orf60
Gene Name
chromosome 5 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142306.2 Intron NP_001135778.1
NM_001305388.1 Intron NP_001292317.1
XM_011534534.2 Intron XP_011532836.1
XM_011534535.1 Intron XP_011532837.1
Gene
HNRNPH1
Gene Name
heterogeneous nuclear ribonucleoprotein H1 (H)
There are no transcripts associated with this gene.

View Full Product Details