Product Details

SNP ID
rs145620447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160563368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTTCCTAGGATTTGGGACTCT[G/T]TAAAAATGAGAAAGTCCCAGGCAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP10B PubMed Links

Gene Details

Gene
ATP10B
Gene Name
ATPase phospholipid transporting 10B (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025153.2 6875 UTR 3 NP_079429.2
XM_006714833.2 6875 UTR 3 XP_006714896.1
XM_011534468.2 6875 UTR 3 XP_011532770.1
XM_011534469.1 6875 UTR 3 XP_011532771.1
XM_011534472.2 6875 UTR 3 XP_011532774.1
XM_017009252.1 6875 UTR 3 XP_016864741.1
XM_017009253.1 6875 UTR 3 XP_016864742.1

View Full Product Details