Product Details

SNP ID
rs145802190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:33945898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTGGTTTTTATTTAAAAATGC[C/T]GGAAAGATTAACAAAAAACTAGTAA
Phenotype
MIM: 609445 MIM: 606202
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RXFP3 PubMed Links

Gene Details

Gene
RXFP3
Gene Name
relaxin/insulin like family peptide receptor 3
There are no transcripts associated with this gene.

Gene
SLC45A2
Gene Name
solute carrier family 45 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012509.3 2725 UTR 3 NP_001012527.1
NM_001297417.2 2725 Intron NP_001284346.2
NM_016180.4 2725 Intron NP_057264.3

View Full Product Details