Product Details
- SNP ID
-
rs146045012
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:177022774 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGCGCCGGCCACGGTGCAGGCCGC[C/G]GACGGCGGAGCGGCCGGGCCTTACA
- Phenotype
-
MIM: 609433
MIM: 605308
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UIMC1
PubMed Links
Gene Details
- Gene
- UIMC1
- Gene Name
- ubiquitin interaction motif containing 1
- Gene
- ZNF346
- Gene Name
- zinc finger protein 346
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308213.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295142.1 |
NM_001308214.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295143.1 |
NM_001308215.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295144.1 |
NM_001308216.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295145.1 |
NM_001308218.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295147.1 |
NM_001308219.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295148.1 |
NM_001308221.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295150.1 |
NM_001308223.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_001295152.1 |
NM_012279.3 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
NP_036411.1 |
XM_005265860.2 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_005265917.1 |
XM_005265864.3 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_005265921.1 |
XM_011534493.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_011532795.1 |
XM_011534494.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_011532796.1 |
XM_011534495.2 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_011532797.1 |
XM_011534496.2 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_011532798.1 |
XM_011534497.2 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_011532799.1 |
XM_017009294.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864783.1 |
XM_017009295.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864784.1 |
XM_017009296.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864785.1 |
XM_017009297.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864786.1 |
XM_017009298.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864787.1 |
XM_017009299.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864788.1 |
XM_017009300.1 |
95 |
Intron |
|
|
XP_016864789.1 |
XM_017009301.1 |
95 |
Silent Mutation |
GCC,GCG |
A12A |
XP_016864790.1 |
XM_017009302.1 |
95 |
UTR 5 |
|
|
XP_016864791.1 |
XM_017009303.1 |
95 |
Intron |
|
|
XP_016864792.1 |
View Full Product Details