Product Details

SNP ID
rs146047826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:168563950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAATCTCCTCCATAAATATCAC[A/G]GACCAGCTTGTCAGCTTGTGTGCTA
Phenotype
MIM: 613187 MIM: 606161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR103A1 PubMed Links

Gene Details

Gene
MIR103A1
Gene Name
microRNA 103a-1
There are no transcripts associated with this gene.

Gene
MIR103B1
Gene Name
microRNA 103b-1
There are no transcripts associated with this gene.

Gene
PANK3
Gene Name
pantothenate kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024594.3 1077 Missense Mutation CGT,TGT R251C NP_078870.1

View Full Product Details