Product Details

SNP ID
rs146445557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127524579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATGTATCCTCCATGGAGTCCT[G/T]CCCACCACTCGCATACTCTACTCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRRC1 PubMed Links

Gene Details

Gene
PRRC1
Gene Name
proline rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286808.1 340 Missense Mutation TGC,TTC C51F NP_001273737.1
NM_130809.4 340 Missense Mutation TGC,TTC C51F NP_570721.1
XM_006714529.3 340 Missense Mutation TGC,TTC C51F XP_006714592.1

View Full Product Details