Product Details

SNP ID
rs146456008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141639580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCAAGAGGCCTCCACTTGTCC[A/G]TCAGGGACGCTCCAAGGAAGGAAAA
Phenotype
MIM: 605166 MIM: 611213
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCHSD1 PubMed Links

Gene Details

Gene
FCHSD1
Gene Name
FCH and double SH3 domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033449.2 1302 UTR 3 NP_258260.1
XM_005268524.4 1302 Intron XP_005268581.1
XM_006714803.3 1302 Intron XP_006714866.1
XM_011537698.2 1302 Intron XP_011536000.1
XM_011537700.2 1302 Intron XP_011536002.1
XM_011537701.2 1302 Intron XP_011536003.1
XM_017010013.1 1302 Intron XP_016865502.1
Gene
HDAC3
Gene Name
histone deacetylase 3
There are no transcripts associated with this gene.

Gene
RELL2
Gene Name
RELT like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130029.1 1302 Missense Mutation CAT,CGT H145R NP_001123501.1
NM_173828.4 1302 Missense Mutation CAT,CGT H145R NP_776189.3
XM_005268414.3 1302 Missense Mutation CAT,CGT H145R XP_005268471.1
XM_011537624.1 1302 Missense Mutation CAT,CGT H145R XP_011535926.1
XM_011537625.2 1302 Missense Mutation CAT,CGT H145R XP_011535927.1

View Full Product Details