Product Details
- SNP ID
-
rs146491803
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:174728295 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCCCCTGACATTTACATTTATGG[A/G]CTAGCTAATTTTTCCCTTCGTCTTA
- Phenotype
-
MIM: 123101
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MSX2
PubMed Links
Gene Details
- Gene
- MSX2
- Gene Name
- msh homeobox 2
View Full Product Details