Product Details

SNP ID
rs146723522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176651897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGAGAACACCTTCCTGCTCAA[A/G]TTTGTGAGTGCTGCCCTCAAGATCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4E1B PubMed Links

Gene Details

Gene
EIF4E1B
Gene Name
eukaryotic translation initiation factor 4E family member 1B
There are no transcripts associated with this gene.

Gene
TSPAN17
Gene Name
tetraspanin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006616.2 511 Silent Mutation AAA,AAG K94K NP_001006617.2
NM_012171.2 511 Silent Mutation AAA,AAG K94K NP_036303.1
NM_130465.4 511 Silent Mutation AAA,AAG K94K NP_569732.2
XM_005265876.3 511 Intron XP_005265933.1

View Full Product Details