Product Details

SNP ID
rs146892491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134763919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTTCTAGCCACTATCGAAAACG[A/C]TCGGCATCCCGGGGTCGCTCTGGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDX46 PubMed Links

Gene Details

Gene
DDX46
Gene Name
DEAD-box helicase 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300860.1 201 Silent Mutation CGA,CGC R11R NP_001287789.1
NM_014829.3 201 Silent Mutation CGA,CGC R11R NP_055644.2
XM_011543758.1 201 Silent Mutation CGA,CGC R11R XP_011542060.1

View Full Product Details