Product Details

SNP ID
rs148368384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:175672898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAGACGGGAATGTCCAGGTCAT[C/T]AGCAGGTCACAGACCAAGCTCTGGG
Phenotype
MIM: 142703
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRH2 PubMed Links

Gene Details

Gene
HRH2
Gene Name
histamine receptor H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131055.1 Intron NP_001124527.1
NM_022304.2 Intron NP_071640.1
XM_005265904.3 Intron XP_005265961.1
XM_006714864.3 Intron XP_006714927.1
XM_006714865.3 Intron XP_006714928.1
XM_011534548.2 Intron XP_011532850.1
XM_011534549.2 Intron XP_011532851.1
XM_017009426.1 Intron XP_016864915.1

View Full Product Details