Product Details

SNP ID
rs148560544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:124641374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGATCCCTTTCCCTTTCTGCCTCC[C/T]GTTCCCGCTCAGCTGTAGCCTTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF608 PubMed Links

Gene Details

Gene
ZNF608
Gene Name
zinc finger protein 608
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020747.2 3699 Missense Mutation NP_065798.2
XM_005272037.2 3699 Missense Mutation XP_005272094.1
XM_005272038.1 3699 Missense Mutation XP_005272095.1
XM_005272039.2 3699 Missense Mutation XP_005272096.1
XM_005272040.3 3699 Missense Mutation XP_005272097.1
XM_011543520.2 3699 Intron XP_011541822.1
XM_011543522.2 3699 Missense Mutation XP_011541824.1
XM_011543523.2 3699 Missense Mutation XP_011541825.1
XM_017009674.1 3699 Missense Mutation XP_016865163.1

View Full Product Details