Product Details

SNP ID
rs148788452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83940697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATATATACAAAGAAAAAAACAAC[A/G]TTGGAATATTACACAGCTTGAAGGT
Phenotype
MIM: 606018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDIL3 PubMed Links

Gene Details

Gene
EDIL3
Gene Name
EGF like repeats and discoidin domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278642.1 4658 UTR 3 NP_001265571.1
NM_005711.4 4658 UTR 3 NP_005702.3

View Full Product Details