Product Details

SNP ID
rs149016123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56909711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTTACAAGTACCGCTTCGTTCC[C/G]TGGATCGCACTGAACCTAAGCCACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIER3 PubMed Links

Gene Details

Gene
MIER3
Gene Name
MIER family member 3
There are no transcripts associated with this gene.

Gene
SETD9
Gene Name
SET domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171990.2 452 Silent Mutation CCC,CCG P22P NP_001165461.1
NM_001323018.1 452 Intron NP_001309947.1
NM_001323019.1 452 Silent Mutation CCC,CCG P22P NP_001309948.1
NM_001323020.1 452 Intron NP_001309949.1
NM_001323022.1 452 Intron NP_001309951.1
NM_153706.3 452 Silent Mutation CCC,CCG P22P NP_714917.2
XM_005248426.4 452 Intron XP_005248483.1
XM_011543139.2 452 Intron XP_011541441.1
XM_011543140.2 452 Intron XP_011541442.1

View Full Product Details