Product Details

SNP ID
rs149065023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180848973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCCTTACCACACTCATTACAT[C/T]GGTATGGCTTCTTTCCAGTGTGGAT
Phenotype
MIM: 610281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP62 PubMed Links

Gene Details

Gene
ZFP62
Gene Name
ZFP62 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172638.1 2692 Missense Mutation CAA,CGA Q841R NP_001166109.1
NM_152283.4 2692 Missense Mutation CAA,CGA Q808R NP_689496.4
XM_017009710.1 2692 Missense Mutation CAA,CGA Q808R XP_016865199.1
XM_017009711.1 2692 Missense Mutation CAA,CGA Q808R XP_016865200.1
XM_017009712.1 2692 Missense Mutation CAA,CGA Q808R XP_016865201.1
XM_017009713.1 2692 Missense Mutation CAA,CGA Q808R XP_016865202.1
XM_017009714.1 2692 Missense Mutation CAA,CGA Q808R XP_016865203.1
XM_017009715.1 2692 Missense Mutation CAA,CGA Q808R XP_016865204.1
XM_017009716.1 2692 Intron XP_016865205.1
XM_017009717.1 2692 Missense Mutation CAA,CGA Q841R XP_016865206.1
XM_017009718.1 2692 Intron XP_016865207.1
XM_017009719.1 2692 Intron XP_016865208.1
XM_017009720.1 2692 Intron XP_016865209.1
XM_017009721.1 2692 Intron XP_016865210.1

View Full Product Details