Product Details

SNP ID
rs149567448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79693909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACAAATATAGCAGGATAAATGG[A/G]TGGGGTATATTTGGATGAGGTAGTG
Phenotype
MIM: 612193 MIM: 614121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMYA5 PubMed Links

Gene Details

Gene
CMYA5
Gene Name
cardiomyopathy associated 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153610.4 Intron NP_705838.3
XM_017009212.1 Intron XP_016864701.1
Gene
PAPD4
Gene Name
PAP associated domain containing 4
There are no transcripts associated with this gene.

View Full Product Details