Product Details

SNP ID
rs149731473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163439417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGACTGCAAGACTAAGGGACT[G/T]TGAAGTAAAAGATCTTCTTAGTCTA
Phenotype
MIM: 601578
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNG1 PubMed Links

Gene Details

Gene
CCNG1
Gene Name
cyclin G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004060.3 395 Missense Mutation TGT,TTT C54F NP_004051.1
NM_199246.1 395 Missense Mutation TGT,TTT C54F NP_954854.1

View Full Product Details