Product Details

SNP ID
rs149820426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134607875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGTAAATCTCTGGGGTTCTTC[C/T]ACTGTTTATCTGGATTTTTTCAATG
Phenotype
MIM: 607690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAR1B PubMed Links

Gene Details

Gene
SAR1B
Gene Name
secretion associated Ras related GTPase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033503.2 Intron NP_001028675.1
NM_016103.3 Intron NP_057187.1
XM_017009521.1 Intron XP_016865010.1

View Full Product Details