Product Details

SNP ID
rs150394007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159917578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGTCATTCTAGTCATGTACTGC[C/G]GTGTCTATATAGTGGCCAAGAGAAC
Phenotype
MIM: 104220
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADRA1B PubMed Links

Gene Details

Gene
ADRA1B
Gene Name
adrenoceptor alpha 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000679.3 846 Missense Mutation CGT,GGT R225G NP_000670.1
XM_005265818.3 846 Missense Mutation CGT,GGT R225G XP_005265875.1
XM_005265819.2 846 Missense Mutation CGT,GGT R225G XP_005265876.1
XM_006714821.3 846 Missense Mutation CGT,GGT R225G XP_006714884.1
XM_011534435.1 846 Missense Mutation CGT,GGT R225G XP_011532737.1
XM_011534437.2 846 Missense Mutation CGT,GGT R225G XP_011532739.1
XM_011534438.2 846 Missense Mutation CGT,GGT R225G XP_011532740.1

View Full Product Details