Product Details

SNP ID
rs150426064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140265744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGCAGCAGGACGATCCCTCTG[C/T]GGAGCCCAGGACATACGGAATGCTG
Phenotype
MIM: 604897
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PFDN1 PubMed Links

Gene Details

Gene
PFDN1
Gene Name
prefoldin subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002622.4 Intron NP_002613.2
XM_005268465.3 Intron XP_005268522.1

View Full Product Details