Product Details

SNP ID
rs150492112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53484153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCTACTGTGTGACCTGTAATC[A/G]GATTTGCCCAGAGCCTGCTTCCTCT
Phenotype
MIM: 136470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FST PubMed Links

Gene Details

Gene
FST
Gene Name
follistatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006350.3 749 Missense Mutation CAG,CGG Q194R NP_006341.1
NM_013409.2 749 Missense Mutation CAG,CGG Q194R NP_037541.1
XM_005248400.4 749 Missense Mutation CAG,CGG Q194R XP_005248457.1
XM_005248401.4 749 Missense Mutation CAG,CGG Q194R XP_005248458.1
XM_005248402.4 749 Missense Mutation CAG,CGG Q194R XP_005248459.1
XM_005248403.4 749 Missense Mutation CAG,CGG Q194R XP_005248460.1
XM_011543099.2 749 Missense Mutation CAG,CGG Q189R XP_011541401.1
XM_017008954.1 749 Missense Mutation CAG,CGG Q194R XP_016864443.1
XM_017008955.1 749 Missense Mutation CAG,CGG Q116R XP_016864444.1

View Full Product Details