Product Details

SNP ID
rs150541427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139420557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACGTTGTGTAGGATCTAGTAT[A/G]GTACCTGACAGAGTAGGCTATTATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC18 PubMed Links

Gene Details

Gene
DNAJC18
Gene Name
DnaJ heat shock protein family (Hsp40) member C18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152686.3 Intron NP_689899.1

View Full Product Details