Product Details

SNP ID
rs151063287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43121978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTGTTGTTGTTTTATGCTT[C/T]AGGGGTTTTGTTCCTCGGCTCGAGC
Phenotype
MIM: 604073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF131 PubMed Links

Gene Details

Gene
ZNF131
Gene Name
zinc finger protein 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248359.2 363 Intron XP_005248416.1
XM_005248360.2 363 Intron XP_005248417.1
XM_005248361.2 363 Intron XP_005248418.1
XM_005248362.4 363 Intron XP_005248419.1
XM_005248363.4 363 Intron XP_005248420.1
XM_005248365.3 363 Intron XP_005248422.1
XM_005248367.2 363 Intron XP_005248424.1
XM_011514123.2 363 Intron XP_011512425.1
XM_011514124.2 363 Intron XP_011512426.1
XM_017009830.1 363 Intron XP_016865319.1
XM_017009831.1 363 Intron XP_016865320.1
XM_017009832.1 363 Intron XP_016865321.1
XM_017009833.1 363 Intron XP_016865322.1
XM_017009834.1 363 Intron XP_016865323.1
XM_017009835.1 363 Intron XP_016865324.1
XM_017009836.1 363 UTR 5 XP_016865325.1
XM_017009837.1 363 UTR 5 XP_016865326.1
XM_017009838.1 363 Intron XP_016865327.1
XM_017009839.1 363 Intron XP_016865328.1
XM_017009840.1 363 Intron XP_016865329.1
XM_017009841.1 363 Intron XP_016865330.1
XM_017009842.1 363 Intron XP_016865331.1
XM_017009843.1 363 Intron XP_016865332.1

View Full Product Details