Product Details
- SNP ID
-
rs151242606
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141095171 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAGGCGGAGCTACGGATTAGGGA[C/T]GTAAATGATCATTCCCCAGTTTTCC
- Phenotype
-
MIM: 606328
MIM: 606329
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs31853] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCDHB2
- Gene Name
- protocadherin beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018936.3 |
565 |
Silent Mutation |
GAC,GAT |
D127D |
NP_061759.1 |
- Gene
- PCDHB3
- Gene Name
- protocadherin beta 3
There are no transcripts associated with this gene.
View Full Product Details